Assay Design



Gene Transcripts

Gene Symbol
LOC101117458
Entrez Gene ID
101117458
Gene Name
UTP11 small subunit processome component
Gene Aliases
UTP11
Chromosome Location
Chr.1: 13243309 - 13254576 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012100981.3 XP_011956371.1 2 - 3 225 81

View Full Product Details