Assay Design



Gene Transcripts

Gene Symbol
VMA21
Entrez Gene ID
101114589
Gene Name
vacuolar ATPase assembly factor VMA21
Gene Aliases
-
Chromosome Location
Chr.X: 91313583 - 91324304 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022279.4 XP_004022328.1 2 - 3 260 105
XM_012107165.3 XP_011962555.1 3 - 4 536 105

View Full Product Details