Assay Design



Gene Transcripts

Gene Symbol
ZNF75D
Entrez Gene ID
101106826
Gene Name
zinc finger protein 75D
Gene Aliases
-
Chromosome Location
Chr.X: 108381781 - 108409832 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012107284.3 XP_011962674.2 2 - 3 798 77
XM_012107287.3 XP_011962677.2 2 - 3 781 77
XM_012107288.3 XP_011962678.2 2 - 3 708 77
XM_012107289.3 XP_011962679.2 2 - 3 708 77
XM_012107291.3 XP_011962681.2 2 - 3 708 77
XM_027963174.1 XP_027818975.1 2 - 3 708 77
XM_027963175.1 XP_027818976.1 2 - 3 708 77
XM_027963176.1 XP_027818977.1 2 - 3 708 77

View Full Product Details