Assay Design



Gene Transcripts

Gene Symbol
SH3RF3
Entrez Gene ID
101109609
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
-
Chromosome Location
Chr.3: 66472031 - 66631819 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966794.1 XP_027822595.1 7 - 8 2736 76

View Full Product Details