Assay Design



Gene Transcripts

Gene Symbol
CENPW
Entrez Gene ID
101117252
Gene Name
centromere protein W
Gene Aliases
-
Chromosome Location
Chr.8: 13705351 - 13714428 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011137.4 XP_004011186.1 1 - 2 307 98

View Full Product Details