Assay Design



Gene Transcripts

Gene Symbol
SUFU
Entrez Gene ID
101114993
Gene Name
SUFU negative regulator of hedgehog signaling
Gene Aliases
-
Chromosome Location
Chr.22: 25733050 - 25843036 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020324.4 XP_004020373.4 2 - 3 467 63
XM_015103389.2 XP_014958875.2 2 - 3 468 63
XM_015103390.2 XP_014958876.2 2 - 3 468 63

View Full Product Details