Assay Design



Gene Transcripts

Gene Symbol
DEF6
Entrez Gene ID
101117371
Gene Name
DEF6 guanine nucleotide exchange factor
Gene Aliases
-
Chromosome Location
Chr.20: 10953351 - 10975441 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018767.4 XP_004018816.2 10 - 11 1697 74
XM_027958605.1 XP_027814406.1 10 - 11 1709 74

View Full Product Details