Assay Design



Gene Transcripts

Gene Symbol
PPP1R14D
Entrez Gene ID
101111024
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
Gene Aliases
-
Chromosome Location
Chr.7: 36349696 - 36371334 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010454.4 XP_004010503.1 1 - 2 440 73
XM_027971691.1 XP_027827492.1 1 - 2 453 73

View Full Product Details