Assay Design



Gene Transcripts

Gene Symbol
EMILIN2
Entrez Gene ID
101123002
Gene Name
elastin microfibril interfacer 2
Gene Aliases
-
Chromosome Location
Chr.23: 42276016 - 42334369 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103646.2 XP_014959132.2 4 - 5 2668 61
XM_027960820.1 XP_027816621.1 3 - 4 2459 61

View Full Product Details