Assay Design



Gene Transcripts

Gene Symbol
STEAP2
Entrez Gene ID
101111960
Gene Name
STEAP2 metalloreductase
Gene Aliases
-
Chromosome Location
Chr.4: 81920208 - 81945285 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007966.3 XP_004008015.1 3 - 4 1277 109

View Full Product Details