Assay Design



Gene Transcripts

Gene Symbol
BLVRB
Entrez Gene ID
101119750
Gene Name
biliverdin reductase B
Gene Aliases
-
Chromosome Location
Chr.14: 52898612 - 52916232 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978310.1 XP_027834111.1 4 - 5 603 76

View Full Product Details