Assay Design



Gene Transcripts

Gene Symbol
MRE11
Entrez Gene ID
101114806
Gene Name
MRE11 homolog, double strand break repair nuclease
Gene Aliases
MRE11A
Chromosome Location
Chr.15: 1165517 - 1240767 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012095281.2 XP_011950671.2 4 - 5 537 69
XM_012095282.3 XP_011950672.2 4 - 5 501 69
XM_012095283.2 XP_011950673.2 2 - 3 163 69

View Full Product Details