Assay Design



Gene Transcripts

Gene Symbol
MRE11
Entrez Gene ID
101114806
Gene Name
MRE11 homolog, double strand break repair nuclease
Gene Aliases
MRE11A
Chromosome Location
Chr.15: 1165517 - 1240767 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012095281.2 XP_011950671.2 6 - 7 764 91
XM_012095282.3 XP_011950672.2 6 - 7 728 91
XM_012095283.2 XP_011950673.2 4 - 5 390 91
XM_012095284.2 XP_011950674.2 3 - 4 318 91

View Full Product Details