Assay Design



Gene Transcripts

Gene Symbol
UBE2S
Entrez Gene ID
101120352
Gene Name
ubiquitin conjugating enzyme E2 S
Gene Aliases
-
Chromosome Location
Chr.14: 66778945 - 66782993 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015419.4 XP_004015468.1 3 - 4 765 92

View Full Product Details