Assay Design



Gene Transcripts

Gene Symbol
SAMD15
Entrez Gene ID
101110598
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
-
Chromosome Location
Chr.7: 92583394 - 92594000 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011085.4 XP_004011134.1 2 - 3 1889 78

View Full Product Details