Assay Design



Gene Transcripts

Gene Symbol
SGCZ
Entrez Gene ID
101122410
Gene Name
sarcoglycan zeta
Gene Aliases
-
Chromosome Location
Chr.26: 23826513 - 25165500 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021739.4 XP_004021788.2 7 - 8 1475 83

View Full Product Details