Assay Design



Gene Transcripts

Gene Symbol
REEP5
Entrez Gene ID
101121779
Gene Name
receptor accessory protein 5
Gene Aliases
-
Chromosome Location
Chr.7: 971691 - 1020967 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096696.2 XP_014952182.2 4 - 5 642 75

View Full Product Details