Assay Design



Gene Transcripts

Gene Symbol
JOSD1
Entrez Gene ID
101105900
Gene Name
Josephin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 231147272 - 231158472 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095023.2 XP_014950509.1 2 - 3 877 88

View Full Product Details