Assay Design



Gene Transcripts

Gene Symbol
CA5A
Entrez Gene ID
101106195
Gene Name
carbonic anhydrase 5A
Gene Aliases
-
Chromosome Location
Chr.14: 14127043 - 14152989 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977721.1 XP_027833522.1 3 - 4 508 67

View Full Product Details