Assay Design



Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
101104182
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
-
Chromosome Location
Chr.18: 18528547 - 18535622 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017748.4 XP_004017797.2 1 - 2 188 91

View Full Product Details