Assay Design



Gene Transcripts

Gene Symbol
IDNK
Entrez Gene ID
101115944
Gene Name
IDNK gluconokinase
Gene Aliases
-
Chromosome Location
Chr.2: 37982362 - 38000022 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093056.2 XP_014948542.1 3 - 4 231 90
XM_015093057.2 XP_014948543.1 3 - 4 182 90
XM_015093058.2 XP_014948544.1 2 - 3 206 90

View Full Product Details