Assay Design



Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
101112250
Gene Name
kallikrein related peptidase 10
Gene Aliases
-
Chromosome Location
Chr.14: 60493089 - 60497988 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015395.3 XP_004015444.2 4 - 5 663 64
XM_012094665.3 XP_011950055.2 4 - 5 665 64

View Full Product Details