Assay Design



Gene Transcripts

Gene Symbol
FUS
Entrez Gene ID
101104799
Gene Name
FUS RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.24: 28033439 - 28043644 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012104405.3 XP_011959795.2 2 - 3 118 65
XM_012104406.3 XP_011959796.2 2 - 3 118 65

View Full Product Details