Assay Design



Gene Transcripts

Gene Symbol
ALG11
Entrez Gene ID
101120844
Gene Name
ALG11 alpha-1,2-mannosyltransferase
Gene Aliases
-
Chromosome Location
Chr.10: 23781287 - 23793930 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012091.4 XP_004012140.1 3 - 4 1227 58
XM_012184513.3 XP_012039903.1 3 - 4 1227 58

View Full Product Details