Assay Design



Gene Transcripts

Gene Symbol
PPM1L
Entrez Gene ID
101105045
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1L
Gene Aliases
-
Chromosome Location
Chr.1: 247061702 - 247393526 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003202.3 XP_004003251.1 2 - 3 1326 96
XM_027958832.1 XP_027814633.1 2 - 3 1849 96

View Full Product Details