Assay Design



Gene Transcripts

Gene Symbol
KDELR1
Entrez Gene ID
101122047
Gene Name
KDEL endoplasmic reticulum protein retention receptor 1
Gene Aliases
-
Chromosome Location
Chr.14: 58536512 - 58544195 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015350.4 XP_004015399.2 2 - 3 393 106

View Full Product Details