Assay Design



Gene Transcripts

Gene Symbol
EIF4E1B
Entrez Gene ID
101106260
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
-
Chromosome Location
Chr.5: 38373790 - 38376770 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970589.1 XP_027826390.1 3 - 4 261 71

View Full Product Details