Assay Design



Gene Transcripts

Gene Symbol
WNT2B
Entrez Gene ID
101116782
Gene Name
Wnt family member 2B
Gene Aliases
-
Chromosome Location
Chr.1: 94675335 - 94690294 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002328.4 XP_004002377.1 3 - 4 931 77

View Full Product Details