Assay Design



Gene Transcripts

Gene Symbol
ASB1
Entrez Gene ID
101108274
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 2718490 - 2742506 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027967064.1 XP_027822865.1 4 - 5 934 66
XM_027967071.1 XP_027822872.1 3 - 4 630 66

View Full Product Details