Assay Design



Gene Transcripts

Gene Symbol
NID1
Entrez Gene ID
101117471
Gene Name
nidogen 1
Gene Aliases
-
Chromosome Location
Chr.25: 8342613 - 8441982 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012105605.3 XP_011960995.3 7 - 8 1852 57
XM_027962362.1 XP_027818163.1 7 - 8 1855 57

View Full Product Details