Assay Design



Gene Transcripts

Gene Symbol
AREG
Entrez Gene ID
443427
Gene Name
amphiregulin
Gene Aliases
-
Chromosome Location
Chr.6: 99250625 - 99260260 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012180163.3 XP_012035553.2 4 - 5 841 73
XM_012180164.3 XP_012035554.2 4 - 5 841 73

View Full Product Details