Assay Design



Gene Transcripts

Gene Symbol
TRAF6
Entrez Gene ID
101107132
Gene Name
TNF receptor associated factor 6
Gene Aliases
-
Chromosome Location
Chr.15: 72104078 - 72124129 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015101111.2 XP_014956597.1 3 - 4 668 92

View Full Product Details