Assay Design



Gene Transcripts

Gene Symbol
KCNK17
Entrez Gene ID
101105697
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
-
Chromosome Location
Chr.20: 14732433 - 14745770 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958491.1 XP_027814292.1 4 - 5 839 85

View Full Product Details