Assay Design



Gene Transcripts

Gene Symbol
LRRC17
Entrez Gene ID
101118781
Gene Name
leucine rich repeat containing 17
Gene Aliases
-
Chromosome Location
Chr.4: 49791673 - 49832102 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007822.4 XP_004007871.1 3 - 4 1428 102

View Full Product Details