Assay Design



Gene Transcripts

Gene Symbol
HYAL4
Entrez Gene ID
101106519
Gene Name
hyaluronidase 4
Gene Aliases
-
Chromosome Location
Chr.4: 96339297 - 96350504 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968381.1 XP_027824182.1 1 - 2 1749 84

View Full Product Details