Assay Design



Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
101115817
Gene Name
natural killer cell granule protein 7
Gene Aliases
-
Chromosome Location
Chr.14: 61170631 - 61172323 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977537.1 XP_027833338.1 2 - 3 549 70

View Full Product Details