Assay Design



Gene Transcripts

Gene Symbol
CRYGN
Entrez Gene ID
101115362
Gene Name
crystallin gamma N
Gene Aliases
-
Chromosome Location
Chr.4: 124044098 - 124049718 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095164.1 XP_014950650.1 1 - 2 22 55

View Full Product Details