Assay Design



Gene Transcripts

Gene Symbol
COL10A1
Entrez Gene ID
101106693
Gene Name
collagen type X alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.8: 24080735 - 24088854 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011185.4 XP_004011234.2 2 - 3 230 70

View Full Product Details