Assay Design



Gene Transcripts

Gene Symbol
LOC105603227
Entrez Gene ID
105603227
Gene Name
serpin A11
Gene Aliases
SERPINA11
Chromosome Location
Chr.18: 59497648 - 59510515 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012098999.3 XP_011954389.2 4 - 5 1143 115

View Full Product Details