Assay Design



Gene Transcripts

Gene Symbol
YY2
Entrez Gene ID
105605393
Gene Name
YY2 transcription factor
Gene Aliases
-
Chromosome Location
Chr.X: 21108369 - 21118998 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963342.1 XP_027819143.1 3 - 3 5830 65

View Full Product Details