Assay Design



Gene Transcripts

Gene Symbol
HPDL
Entrez Gene ID
101119084
Gene Name
4-hydroxyphenylpyruvate dioxygenase like
Gene Aliases
-
Chromosome Location
Chr.1: 20628176 - 20630295 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004001909.4 XP_004001958.1 1 - 1 1737 74

View Full Product Details