Assay Design



Gene Transcripts

Gene Symbol
MIF4GD
Entrez Gene ID
101121439
Gene Name
MIF4G domain containing
Gene Aliases
-
Chromosome Location
Chr.11: 7042841 - 7047285 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974065.1 XP_027829866.1 4 - 5 496 107
XM_027974066.1 XP_027829867.1 4 - 5 482 107

View Full Product Details