Assay Design



Gene Transcripts

Gene Symbol
CCM2
Entrez Gene ID
101114100
Gene Name
CCM2 scaffold protein
Gene Aliases
-
Chromosome Location
Chr.4: 84126473 - 84178536 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968694.1 XP_027824495.1 7 - 8 1009 118
XM_027968695.1 XP_027824496.1 9 - 10 1145 118
XM_027968696.1 XP_027824497.1 10 - 11 1202 118
XM_027968697.1 XP_027824498.1 9 - 10 1140 118
XM_027968698.1 XP_027824499.1 8 - 9 1052 118
XM_027968699.1 XP_027824500.1 6 - 7 931 118
XM_027968700.1 XP_027824501.1 6 - 7 816 118
XM_027968702.1 XP_027824503.1 5 - 6 699 118

View Full Product Details