Assay Design



Gene Transcripts

Gene Symbol
VSIG10L
Entrez Gene ID
101122129
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
-
Chromosome Location
Chr.14: 61136978 - 61151169 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977539.1 XP_027833340.1 2 - 3 1063 66

View Full Product Details