Assay Design



Gene Transcripts

Gene Symbol
LOC101103947
Entrez Gene ID
101103947
Gene Name
alpha-1-acid glycoprotein
Gene Aliases
ORM1
Chromosome Location
Chr.2: 9742547 - 9745957 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003982.4 XP_004004031.1 4 - 5 475 114

View Full Product Details