Assay Design



Gene Transcripts

Gene Symbol
TCTA
Entrez Gene ID
101104532
Gene Name
T cell leukemia translocation altered
Gene Aliases
-
Chromosome Location
Chr.19: 52680221 - 52683330 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018461.4 XP_004018510.1 2 - 3 505 93

View Full Product Details