Assay Design



Gene Transcripts

Gene Symbol
TBCA
Entrez Gene ID
101107793
Gene Name
tubulin folding cofactor A
Gene Aliases
-
Chromosome Location
Chr.7: 9630806 - 9714729 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096795.2 XP_014952281.1 3 - 4 332 92

View Full Product Details