Assay Design



Gene Transcripts

Gene Symbol
DUXA
Entrez Gene ID
101113006
Gene Name
double homeobox A
Gene Aliases
-
Chromosome Location
Chr.14: 69508117 - 69525761 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978012.1 XP_027833813.1 3 - 4 535 64

View Full Product Details