Assay Design



Gene Transcripts

Gene Symbol
RMI2
Entrez Gene ID
101107838
Gene Name
RecQ mediated genome instability 2
Gene Aliases
-
Chromosome Location
Chr.24: 10241781 - 10247188 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021173.4 XP_004021222.3 1 - 2 306 111

View Full Product Details