Assay Design



Gene Transcripts

Gene Symbol
NR0B2
Entrez Gene ID
101114500
Gene Name
nuclear receptor subfamily 0 group B member 2
Gene Aliases
-
Chromosome Location
Chr.2: 255823479 - 255825664 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005496.3 XP_004005545.2 1 - 2 639 59

View Full Product Details